Assay Details
| Gene Symbol: LOC102180424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972083.2 | XP_013827537.2 | 7 - 8 | 2205 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102180424| Gene Symbol: LOC102180424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972083.2 | XP_013827537.2 | 7 - 8 | 2205 | 97 | |
Set Membership: |
|