Assay Details
| Gene Symbol: SKAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064939.1 | XP_017920428.1 | 7 - 8 | 668 | 65 | |
| GenBank mRNA | JO580791.1 | - | 6 - 7 | 944 | 65 | |
More Information
Set Membership: |
|

SKAP1
Genomic Map