Assay Details
| Gene Symbol: SLC22A23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038802.1 | XP_017894291.1 | 5 - 6 | 1211 | 61 | |
| XM_018038803.1 | XP_017894292.1 | 5 - 6 | 1196 | 61 | ||
| XM_018038804.1 | XP_017894293.1 | 5 - 6 | 1196 | 61 | ||
More Information
Set Membership: |
|

SLC22A23
Genomic Map