Assay Details
| Gene Symbol: SRPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043937.1 | XP_017899426.1 | 3 - 4 | 456 | 55 | |
| XM_018043938.1 | XP_017899427.1 | 2 - 3 | 395 | 55 | ||
More Information
Set Membership: |
|

SRPX
Genomic Map