Assay Details
| Gene Symbol: BGLAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046289.1 | XP_017901778.1 | 3 - 4 | 1565 | 79 | |
| GenBank mRNA | AY661470.1 | - | 1 - 2 | 76 | 79 | |
More Information
Set Membership: |
|

BGLAP
Genomic Map