Assay Details
| Gene Symbol: RABL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674990.2 | XP_005675047.1 | 1 - 2 | 104 | 70 | |
| GenBank mRNA | JO367433.2 | - | 6 - 7 | 609 | 70 | |
| JO367434.2 | - | 5 - 6 | 479 | 70 | ||
| JO370163.2 | - | 1 - 2 | 44 | 70 | ||
| JO370164.2 | - | 1 - 2 | 44 | 70 | ||
| JO370165.2 | - | 1 - 2 | 44 | 70 | ||
| JO414699.2 | - | 1 - 2 | 60 | 70 | ||
| JO444415.2 | - | 7 - 8 | 832 | 70 | ||
| JO506692.2 | - | 7 - 8 | 914 | 70 | ||
| JO586191.2 | - | 7 - 8 | 854 | 70 | ||
| JO637969.2 | - | 1 - 2 | 56 | 70 | ||
| JT737672.1 | - | 9 - 10 | 1245 | 70 | ||
| KA325401.1 | - | 1 - 2 | 66 | 70 | ||
More Information
Set Membership: |
|

RABL3
Genomic Map