Assay Details
| Gene Symbol: UAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046423.1 | XP_017901912.1 | 6 - 7 | 1689 | 77 | |
| XM_018046424.1 | XP_017901913.1 | 7 - 8 | 1761 | 77 | ||
| XM_018046425.1 | XP_017901914.1 | 6 - 7 | 1689 | 77 | ||
| XM_018046426.1 | XP_017901915.1 | 6 - 7 | 1689 | 77 | ||
| GenBank mRNA | JO577315.1 | - | 6 - 7 | 1312 | 77 | |
More Information
Set Membership: |
|

UAP1
Genomic Map