Assay Details
| Gene Symbol: C3H1orf162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677896.3 | XP_005677953.1 | 3 - 4 | 394 | 97 | |
| XM_013962544.2 | XP_013817998.1 | 3 - 4 | 560 | 97 | ||
| GenBank mRNA | JO340517.2 | - | 3 - 4 | 338 | 97 | |
More Information
Set Membership: |
|

C3H1orf162
Genomic Map