Assay Details
| Gene Symbol: C3H1orf162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677896.3 | XP_005677953.1 | 4 - 5 | 495 | 96 | |
| GenBank mRNA | JO340517.2 | - | 4 - 5 | 439 | 96 | |
More Information
Set Membership: |
|

C3H1orf162
Genomic Map