Assay Details
| Gene Symbol: C3H1orf162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677896.3 | XP_005677953.1 | 5 - 6 | 540 | 79 | |
| XM_013962544.2 | XP_013817998.1 | 5 - 6 | 750 | 79 | ||
| GenBank mRNA | JO340517.2 | - | 5 - 6 | 484 | 79 | |
| JT686739.1 | - | 2 - 3 | 572 | 79 | ||
More Information
Set Membership: |
|

C3H1orf162
Genomic Map