Assay Details
| Gene Symbol: UBE2H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679467.3 | XP_005679524.1 | 4 - 5 | 184 | 105 | |
| XM_018046929.1 | XP_017902418.1 | 5 - 6 | 770 | 105 | ||
More Information
Set Membership: |
|

UBE2H
Genomic Map