Assay Details
| Gene Symbol: SPARCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681882.3 | XP_005681939.2 | 2 - 3 | 759 | 67 | |
| XM_013964780.2 | XP_013820234.2 | 2 - 3 | 84 | 67 | ||
| GenBank mRNA | JO581145.1 | - | 9 - 10 | 2477 | 67 | |
More Information
Set Membership: |
|

SPARCL1
Genomic Map