Assay Details
| Gene Symbol: POGLUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687733.3 | XP_005687790.1 | 5 - 6 | 1302 | 74 | |
| GenBank mRNA | KA334560.1 | - | 5 - 6 | 812 | 74 | |
More Information
Set Membership: |
|

POGLUT2
Genomic Map