Assay Details
| Gene Symbol: POGLUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687733.3 | XP_005687790.1 | 6 - 7 | 1533 | 70 | |
| GenBank mRNA | KA334560.1 | - | 4 - 5 | 581 | 70 | |
More Information
Set Membership: |
|

POGLUT2
Genomic Map