Assay Details
| Gene Symbol: IL18BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689870.3 | XP_005689927.1 | 2 - 3 | 281 | 75 | |
| XM_018059420.1 | XP_017914909.1 | 2 - 3 | 281 | 75 | ||
| GenBank mRNA | JO498841.2 | - | 2 - 3 | 1201 | 75 | |
More Information
Set Membership: |
|

IL18BP
Genomic Map