Assay Details
| Gene Symbol: ANGPTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690918.3 | XP_005690975.1 | 2 - 3 | 1324 | 79 | |
| GenBank mRNA | JT709387.1 | - | 4 - 5 | 842 | 79 | |
More Information
Set Membership: |
|

ANGPTL1
Genomic Map