Assay Details
| Gene Symbol: HSCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691708.3 | XP_005691765.3 | 2 - 3 | 395 | 75 | |
| XM_018061048.1 | XP_017916537.1 | 2 - 3 | 396 | 75 | ||
| XM_018061049.1 | XP_017916538.1 | 1 - 2 | 95 | 75 | ||
| GenBank mRNA | KA349629.1 | - | 2 - 3 | 391 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018061049.1 transcripts.
Set Membership: |
|

HSCB
Genomic Map