Assay Details
| Gene Symbol: PSMC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692406.3 | XP_005692463.1 | 2 - 3 | 227 | 58 | |
| GenBank mRNA | JO412915.2 | - | 2 - 3 | 326 | 58 | |
More Information
Set Membership: |
|

PSMC4
Genomic Map