Assay Details
| Gene Symbol: LOC102185494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062528.1 | XP_017918017.1 | 4 | 566 | 127 | |
| XM_018062529.1 | XP_017918018.1 | 4 | 567 | 127 | ||
| GenBank mRNA | JO620528.2 | - | 3 | 407 | 127 | |
| JO620529.2 | - | 3 | 407 | 127 | ||
More Information
Set Membership: |
|

LOC102185494
Genomic Map