Assay Details
| Gene Symbol: MRPS24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018067017.1 | XP_017922506.1 | 3 - 4 | 360 | 63 | |
| GenBank mRNA | JO610784.2 | - | 3 - 4 | 232 | 63 | |
| JT680277.1 | - | 3 - 4 | 206 | 63 | ||
More Information
Set Membership: |
|

MRPS24
Genomic Map