Assay Details
| Gene Symbol: EXOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038297.1 | XP_017893786.1 | 3 - 4 | 514 | 77 | |
| XM_018038298.1 | XP_017893787.1 | 3 - 4 | 514 | 77 | ||
| GenBank mRNA | JO438195.2 | - | 3 - 4 | 490 | 77 | |
More Information
Set Membership: |
|

EXOG
Genomic Map