Assay Details
| Gene Symbol: LOC102183469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696368.3 | XP_005696425.2 | 11 - 12 | 1622 | 120 | |
| XM_013973925.2 | XP_013829379.2 | 10 - 11 | 1138 | 120 | ||
More Information
Set Membership: |
|

LOC102183469
Genomic Map