Assay Details
| Gene Symbol: LOC102183469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696368.3 | XP_005696425.2 | 5 - 6 | 1012 | 97 | |
| XM_013973925.2 | XP_013829379.2 | 4 - 5 | 528 | 97 | ||
| GenBank mRNA | JO434721.2 | - | 6 - 7 | 547 | 97 | |
More Information
Set Membership: |
|

LOC102183469
Genomic Map