Assay Details
| Gene Symbol: CDIPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040956.1 | XP_017896445.1 | 4 - 5 | 811 | 56 | |
| GenBank mRNA | JI861694.1 | - | 4 - 5 | 411 | 56 | |
| JO430990.2 | - | 4 - 5 | 800 | 56 | ||
| JO577346.1 | - | 4 - 5 | 859 | 56 | ||
More Information
Set Membership: |
|

CDIPT
Genomic Map