Assay Details
| Gene Symbol: PDLIM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698725.3 | XP_005698782.1 | 7 - 8 | 998 | 65 | |
| XM_005698726.3 | XP_005698783.1 | 6 - 7 | 854 | 65 | ||
| XM_005698727.3 | XP_005698784.1 | 6 - 7 | 733 | 65 | ||
| GenBank mRNA | JI861762.1 | - | 6 - 7 | 760 | 65 | |
| JO538681.2 | - | 1 - 2 | 949 | 65 | ||
More Information
Set Membership: |
|

PDLIM3
Genomic Map