Assay Details
| Gene Symbol: ZNF248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042274.1 | XP_017897763.1 | 4 - 5 | 353 | 82 | |
| XM_018042275.1 | XP_017897764.1 | 4 - 5 | 410 | 82 | ||
More Information
Set Membership: |
|

ZNF248
Genomic Map