Assay Details
| Gene Symbol: TSPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048872.1 | XP_017904361.1 | 2 - 3 | 313 | 82 | |
| XM_018048873.1 | XP_017904362.1 | 2 - 3 | 223 | 82 | ||
| XM_018048874.1 | XP_017904363.1 | 2 - 3 | 312 | 82 | ||
| GenBank mRNA | JO579273.1 | - | 2 - 3 | 324 | 82 | |
| JO619789.2 | - | 2 - 3 | 336 | 82 | ||
More Information
Set Membership: |
|

TSPO
Genomic Map