Assay Details
| Gene Symbol: ROR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052509.1 | XP_017907998.1 | 4 - 5 | 1342 | 63 | |
| GenBank mRNA | KA336933.1 | - | 5 - 6 | 2905 | 63 | |
More Information
Set Membership: |
|

ROR2
Genomic Map