Assay Details
| Gene Symbol: OVOL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057232.1 | XP_017912721.1 | 3 - 4 | 2374 | 70 | |
| GenBank mRNA | KA328538.1 | - | 1 - 2 | 48 | 70 | |
More Information
Set Membership: |
|

OVOL2
Genomic Map