Assay Details
| Gene Symbol: CCN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968914.2 | XP_013824368.2 | 3 - 4 | 932 | 118 | |
| XM_018057907.1 | XP_017913396.1 | 2 - 3 | 1073 | 118 | ||
| GenBank mRNA | KA312828.1 | - | 2 - 3 | 392 | 118 | |
More Information
Set Membership: |
|

CCN5
Genomic Map