Assay Details
| Gene Symbol: CPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058257.1 | XP_017913746.1 | 4 - 5 | 1031 | 61 | |
| XM_018058258.1 | XP_017913747.1 | 4 - 5 | 937 | 61 | ||
| GenBank mRNA | JO429287.2 | - | 4 - 5 | 1256 | 61 | |
| KA308047.1 | - | 4 - 5 | 1136 | 61 | ||
More Information
Set Membership: |
|

CPQ
Genomic Map