Assay Details
| Gene Symbol: CTSO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061204.1 | XP_017916693.1 | 8 - 9 | 1072 | 64 | |
| GenBank mRNA | JO405320.2 | - | 5 - 6 | 687 | 64 | |
| KA351649.1 | - | 1 - 2 | 668 | 64 | ||
More Information
Set Membership: |
|

CTSO
Genomic Map