Assay Details
| Gene Symbol: AKT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062407.1 | XP_017917896.1 | 9 - 10 | 1096 | 68 | |
| XM_018062408.1 | XP_017917897.1 | 9 - 10 | 1015 | 68 | ||
| XM_018062409.1 | XP_017917898.1 | 9 - 10 | 939 | 68 | ||
| XM_018062410.1 | XP_017917899.1 | 9 - 10 | 1325 | 68 | ||
| GenBank mRNA | JO421234.2 | - | 9 - 10 | 947 | 68 | |
More Information
Set Membership: |
|

AKT2
Genomic Map