Assay Details
| Gene Symbol: LIPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062482.1 | XP_017917971.1 | 4 - 5 | 956 | 61 | |
| XM_018062483.1 | XP_017917972.1 | 4 - 5 | 820 | 61 | ||
| XM_018062484.1 | XP_017917973.1 | 4 - 5 | 1058 | 61 | ||
| GenBank mRNA | EU273879.1 | - | 3 - 4 | 751 | 61 | |
| GQ927175.1 | - | 3 - 4 | 643 | 61 | ||
More Information
Set Membership: |
|

LIPE
Genomic Map