Assay Details
| Gene Symbol: LOC102185508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065917.1 | XP_017921406.1 | 6 - 7 | 618 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102185508| Gene Symbol: LOC102185508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065917.1 | XP_017921406.1 | 6 - 7 | 618 | 66 | |
Set Membership: |
|