Assay Details
| Gene Symbol: SLC6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038407.1 | XP_017893896.1 | 6 - 7 | 855 | 54 | |
| XM_018038408.1 | XP_017893897.1 | 5 - 6 | 1279 | 54 | ||
| GenBank mRNA | JO632130.2 | - | 6 - 7 | 922 | 54 | |
More Information
Set Membership: |
|

SLC6A1
Genomic Map