Assay Details
| Gene Symbol: JMJD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039975.1 | XP_017895464.1 | 8 - 9 | 837 | 58 | |
| XM_018039976.1 | XP_017895465.1 | 7 - 8 | 738 | 58 | ||
| GenBank mRNA | JO429465.2 | - | 8 - 9 | 744 | 58 | |
More Information
Set Membership: |
|

JMJD8
Genomic Map