Assay Details
| Gene Symbol: GNPTG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040420.1 | XP_017895909.1 | 9 - 10 | 771 | 79 | |
| XM_018040422.1 | XP_017895911.1 | 8 - 9 | 670 | 79 | ||
| GenBank mRNA | KA309732.1 | - | 9 - 10 | 771 | 79 | |
More Information
Set Membership: |
|

GNPTG
Genomic Map