Assay Details
| Gene Symbol: GPR87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675440.3 | XP_005675497.1 | 2 - 3 | 431 | 85 | |
| XM_018049012.1 | XP_017904501.1 | 3 - 4 | 342 | 85 | ||
| GenBank mRNA | KA304723.1 | - | 1 - 2 | 1795 | 85 | |
More Information
Set Membership: |
|

GPR87
Genomic Map