Assay Details
| Gene Symbol: FAM78B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677089.2 | XP_005677146.2 | 1 - 2 | 810 | 66 | |
| GenBank mRNA | JT645237.1 | - | 1 - 2 | 311 | 66 | |
More Information
Set Membership: |
|

FAM78B
Genomic Map