Assay Details
| Gene Symbol: BCL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678157.3 | XP_005678214.1 | 1 - 2 | 328 | 99 | |
| GenBank mRNA | JO514477.2 | - | 1 - 2 | 361 | 99 | |
More Information
Set Membership: |
|

BCL10
Genomic Map