Assay Details
| Gene Symbol: TAFA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680252.3 | XP_005680309.1 | 4 - 5 | 527 | 93 | |
| GenBank mRNA | JO628761.2 | - | 1 - 2 | 224 | 93 | |
More Information
Set Membership: |
|

TAFA2
Genomic Map