Assay Details
| Gene Symbol: PMM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048824.1 | XP_017904313.1 | 7 - 8 | 986 | 65 | |
| GenBank mRNA | JO632841.2 | - | 1 - 2 | 567 | 65 | |
| KA341168.1 | - | 1 - 2 | 645 | 65 | ||
More Information
Set Membership: |
|

PMM1
Genomic Map