Assay Details
| Gene Symbol: CDKN2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051064.1 | XP_017906553.1 | 2 - 3 | 1028 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDKN2D| Gene Symbol: CDKN2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051064.1 | XP_017906553.1 | 2 - 3 | 1028 | 54 | |
Set Membership: |
|