Assay Details
| Gene Symbol: LOC102189950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052093.1 | XP_017907582.1 | 4 - 5 | 827 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102189950| Gene Symbol: LOC102189950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052093.1 | XP_017907582.1 | 4 - 5 | 827 | 103 | |
Set Membership: |
|