Assay Details
| Gene Symbol: SHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683946.3 | XP_005684003.3 | 4 - 5 | 1837 | 58 | |
| XM_018052197.1 | XP_017907686.1 | 4 - 5 | 1837 | 58 | ||
| GenBank mRNA | JT649362.1 | - | 3 - 4 | 810 | 58 | |
| KA335821.1 | - | 4 - 5 | 1336 | 58 | ||
More Information
Set Membership: |
|

SHB
Genomic Map