Assay Details
| Gene Symbol: RTRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054344.1 | XP_017909833.1 | 2 - 3 | 349 | 95 | |
| GenBank mRNA | JO506659.2 | - | 2 - 3 | 286 | 95 | |
| JT718261.1 | - | 2 - 3 | 188 | 95 | ||
More Information
Set Membership: |
|

RTRAF
Genomic Map