Assay Details
| Gene Symbol: RTRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054344.1 | XP_017909833.1 | 4 - 5 | 544 | 72 | |
| GenBank mRNA | JO506659.2 | - | 4 - 5 | 481 | 72 | |
More Information
Set Membership: |
|

RTRAF
Genomic Map