Assay Details
| Gene Symbol: RNF139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688857.3 | XP_005688914.2 | 1 - 2 | 511 | 74 | |
| GenBank mRNA | JO623193.2 | - | 1 - 2 | 1027 | 74 | |
| JT742675.1 | - | 3 - 4 | 2028 | 74 | ||
More Information
Set Membership: |
|

RNF139
Genomic Map