Assay Details
| Gene Symbol: LOC102184087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693242.2 | XP_005693299.1 | 10 - 11 | 1005 | 63 | |
| XM_005693243.2 | XP_005693300.1 | 9 - 10 | 907 | 63 | ||
| GenBank mRNA | JO580163.1 | - | 8 - 9 | 716 | 63 | |
More Information
Set Membership: |
|

LOC102184087
Genomic Map